missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-4-fluorobenzene, 97+%, Thermo Scientific Chemicals

Product Code. 15469117
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 332-48-9
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD00044739
InChI Key JXSPKRUNMHMICQ-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-4-fluorobenzene, 4-fluorophenoxy-ethylbromide, 2-bromoethyl 4-fluorophenyl ether, 4-fluorophenoxyethylbromide, 4-2-bromoethoxy fluorobenzene, 4-fluorophenoxyethyl bromide, beta-bromo-4-fluorophenetole, 2-4-fluorophenoxy ethyl bromide, benzene, 1-2-bromoethoxy-4-fluoro, p-fluorophenoxy ethylbromide
PubChem CID 2064171
IUPAC Name 1-(2-bromoethoxy)-4-fluorobenzene
SMILES FC1=CC=C(OCCBr)C=C1

Spécification

Quantity 250 mg
Formula Weight 219.05
Percent Purity ≥97%
Chemical Name or Material 1-(2-Bromoethoxy)-4-fluorobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'