missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5,6-Hexabromocyclohexane, Thermo Scientific Chemicals

Product Code. 15463559
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation

Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1837-91-8
Molecular Formula C6H6Br6
Molecular Weight (g/mol) 557.54
MDL Number MFCD00059127
InChI Key QFQZKISCBJKVHI-UHFFFAOYSA-N
Synonym benzene hexabromide, cyclohexane, 1,2,3,4,5,6-hexabromo, jak2 inhibitor ii, acmc-1bqjt, trans-alpha-benzene hexabromide, cyclohexane,2,3,4,5,6-hexabromo, 1,2,3,4,5,6-hexabromo-cyclohexane, #, 1,2,3,4,5,6-hexakis bromanyl cyclohexane, 1,2,3,4,5,6-hexabromocyclohexane hplc
PubChem CID 74603
IUPAC Name 1,2,3,4,5,6-hexabromocyclohexane
SMILES BrC1C(Br)C(Br)C(Br)C(Br)C1Br

Specifications

Color White
Quantity 5 g
Formula Weight 557.5
Physical Form Solid
Chemical Name or Material 1,2,3,4,5,6-Hexabromocyclohexane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'