missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-4-iodobutane, 98+%, stab. with copper, Thermo Scientific Chemicals

£34.67 - £164.82

Chemical Identifiers

CAS 461-17-6
Molecular Formula C4H6F3I
Molecular Weight (g/mol) 237.992
MDL Number MFCD00041543
InChI Key LNDGACQEAYKNOI-UHFFFAOYSA-N
Synonym 1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
PubChem CID 2775189
IUPAC Name 1,1,1-trifluoro-4-iodobutane
SMILES C(CC(F)(F)F)CI
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11400278
SDS View Documents
Thermo Scientific Alfa Aesar
B23811.03
1 g
£34.67
1g
Please to purchase this item. Need a web account? Register with us today!
11410278
SDS View Documents
Thermo Scientific Alfa Aesar
B23811.06
5 g
£164.82
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

461-17-6
237.992
LNDGACQEAYKNOI-UHFFFAOYSA-N
2775189
C(CC(F)(F)F)CI
C4H6F3I
MFCD00041543
1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
1,1,1-trifluoro-4-iodobutane

Specifications

461-17-6
126°C to 127°C
C4H6F3I
1.4335
1736064
1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
C(CC(F)(F)F)CI
237.992
237.99
1,1,1-Trifluoro-4-iodobutane, stabilized with copper
1.85
>100°C (212°F)
CF3(CH2)3I
MFCD00041543
Light sensitive
LNDGACQEAYKNOI-UHFFFAOYSA-N
1,1,1-trifluoro-4-iodobutane
2775189
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.