missing translation for 'onlineSavingsMsg'
Learn More

ZO-1 Rabbit anti-Canine, Hamster, Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21773-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881365

  • £339.00 / 150µL
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
Specifications

Specifications

ZO-1
Polyclonal
Unconjugated
TJP1
Tight junction protein 1, Tight junction protein ZO 1, TJP1, ZO 1, ZO1, ZO-1, Zona occludens protein 1, Zonula occludens protein 1
Rabbit
Antigen Affinity Chromatography
RUO
100773110, 21872, 292994, 403752, 7082
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O97758, P39447, Q07157
TJP1
ZO-1 Fusion Protein Ag16454
150 μL
Primary
Canine, Hamster, Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers