missing translation for 'onlineSavingsMsg'
Learn More

Assorted pH Buffers Case, YSI™

Various pH buffers for pH probe calibration.

Brand:  YSI Yellow Spring Instrument 603824

Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 12973347

  • £73.55 / Pack of 6
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Contains two bottles each of pH 4, 7, and 10 calibration buffers (Cat. Nos. 3821, 3822, and 3823)
  • Plastic bottles (475mL/ea.)

Expiration Dates

  • Unopended: 2 years
  • Opened: 6 months
Specifications

Specifications

4, 7, 10
1 Kit (2 bottles ea. 3821,3822 and 3823)
pH Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers