Learn More
WSB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562650
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LVTASYDTNV IMWDPYTGER LRSLHHTQVD PAMDDSDVHI SSLRSVCFSP EGLYLATVAD DRLL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.
This gene encodes a member of the WD-protein subfamily. The encoded protein contains five WD-repeats spanning most of the protein and an SOCS box in the C-terminus. The SOCS box may act as a bridge between specific substrate-binding domains and E3 ubiquitin protein ligases.Specifications
WSB2 | |
Polyclonal | |
Unconjugated | |
WSB2 | |
AA673511; CS box-containing WD protein; SBA2; SOCS box-containing WD protein SWiP-2; Swip2; WD repeat and SOCS box containing 2; WD repeat and SOCS box containing protein 2; WD repeat and SOCS box-containing 2; WD repeat and SOCS box-containing protein 2; WD SOCS-box protein 2; WD-40-repeat-containing protein with a SOCS box 2; Wsb2; WSB-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55884 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYS7 | |
WSB2 | |
Recombinant protein corresponding to Human WSB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only