missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRCH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562018
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LREDVKVLIE LDKCKEKPVP EEAA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a member of the Rho family of GTPases. This protein can activate PAK1 and JNK1, and can induce filopodium formation and stress fiber dissolution. It may also mediate the effects of WNT1 signaling in the regulation of cell morphology, cytoskeletal organization, and cell proliferation. A non-coding transcript variant of this gene results from naturally occurring read-through transcription between this locus and the neighboring DUSP5P (dual specificity phosphatase 5 pseudogene) locus.Specifications
WRCH1 | |
Polyclonal | |
Unconjugated | |
Rhou | |
2310026M05Rik; AI182090; ARHU; CDC42L1; CDC42-like GTPase 1; G28K; GTP-binding protein like 1; GTP-binding protein SB128; GTP-binding protein-like 1; hG28K; LOC678766; LOW QUALITY PROTEIN: rho-related GTP-binding protein RhoU; mG28K; ras homolog family member U; ras homolog gene family, member U; ras-like gene family member U; Ras-like protein member U; rho GTPase-like protein ARHU; Rho-related GTP-binding protein RhoU; rho-related GTP-binding protein RhoU-like; RHOU; Ryu GTPase; SB128; similar to ras homolog gene family, member U; Wnt1 responsive Cdc42 homolog; wnt-1 responsive Cdc42 homolog 1; WRCH1; WRCH-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L0Q8 | |
Rhou | |
Recombinant protein corresponding to Human WRCH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only