missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNT8B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551341
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total WNT8B / Wnt 8b protein.
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 95%, 86% and 71% amino acid identity to the mouse, zebrafish and Xenopus Wnt8B proteins, respectively. The expression patterns of the human and mouse genes appear identical and are restricted to the developing brain. The chromosomal location of this gene to 10q24 suggests it as a candidate gene for partial epilepsy.Specifications
WNT8B | |
Polyclonal | |
Unconjugated | |
WNT8B | |
protein Wnt-8b; wingless related MMTV integration site 8b; wingless-type MMTV integration site family member 8B; wingless-type MMTV integration site family, member 8B; WNT 8b; Wnt family member 8B; WNT8B; wnt-8b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7479 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q93098 | |
WNT8B | |
Synthetic peptide of human WNT8B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |