missing translation for 'onlineSavingsMsg'
Learn More

Wijs solution, Volumetric, 0.1 M ICl, Honeywell Fluka™

Product Code. 15624790
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L

Volumetric, 0.1 M ICl

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7, 7790-99-0
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 24640
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Quantity 4 x 2.5 L
Linear Formula ICl
MDL Number MFCD00036152
UN Number UN2920
Flash Point 40°C (104°F)
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
Formula Weight 162.36
Chemical Name or Material Wijs solution
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Wijs solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, iodinemonochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'