missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers

Unique cone liner design offers superior torque retention and excellent sealing characteristics

£63.45 - £104.00

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ container
View More Specs

Products 6
Product Code Brand Neck Finish Price Quantity & Availability  
Product Code Brand Neck Finish Price Quantity & Availability  
11728839
View Documents Promotion Details
DWK Life Sciences
239250
15-425
£66.30
Pack of 144
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
 
10123793
View Documents Promotion Details
DWK Life Sciences
239253
20-400
£63.45
Pack of 144
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
11754199
View Documents Promotion Details
DWK Life Sciences
239260
33-400
£104.00
Pack of 144
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
11734219
View Documents Promotion Details
DWK Life Sciences
239257
24-400
£70.95
Pack of 144
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
11714189
View Documents Promotion Details
DWK Life Sciences
239259
28-400
£80.95
Pack of 144
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
11704189
View Documents Promotion Details
DWK Life Sciences
239255
22-400
£69.00
Pack of 144
Estimated Shipment: 04-02-2025
to see stock.
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

  • Poly-Seal liner is manufactured from low density polyethylene
  • Phenolic
  • Screw cap
  • Color: Black

Spezifikation

Spezifikation

Black
144 Cs.
Redi-pak™ container
Phenolic Resin
ISO 9001
Polyseal
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate