missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Leak-Resistant Amber HDPE Bottles with Caps

Catalog Number p-44097
Click to view available options
Capacity:
0.1 oz., 4mL
0.3 oz., 8 mL
0.5 oz., 15 mL
1 oz., 30mL
17 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4.2 oz., 125 mL
8.45 oz., 250 mL
Height (Metric):
38 mm
43 mm
56 mm
69 mm
84 mm
101 mm
129 mm
171 mm
210 mm
Neck Finish:
13-425
20-410
24-410
28-410
38-430
Height (English):
1.4 in.
1.69 in.
2.2 in.
2.71 in.
3.3 in.
3.9 in.
5.07 in.
6.73 in.
8.2 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal primary packaging solution for assays in diagnostic kits, and also chemical and storage purposes. With double-seal closure with cap attached.

With double-seal closure with cap attached

Ideal for short- and long-term storage and packaging.

  • Narrow-mouth
  • No drip pour lip for clean pouring
  • Polypropylene cap attached remains tight over an extended time
  • Convenience packed

Capacity 0.1 oz., 4mL
Capacity (Metric) 4 mL
Capacity (English) 0.1 oz.
Color Amber
Diameter (English) 0.62 in.
Material HDPE
Neck Finish 13-425
Certifications/Compliance ISO 9001
Height (Metric) 38 mm
Height (English) 1.4 in.
Closure Material Polypropylene
Closure Size 13-425 mm
Diameter (Metric) 16 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'