missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper
241.30 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Description
- Amber borosilicate glass bottle with narrow mouth
- Protects light-sensitive reagents and solutions
- Ground-glass octagon flat stopper provides excellent seal; protects top lip of bottle from dust
Specifications
Capacity | 3.38 oz., 100mL |
Capacity (Metric) | 100 mL |
Capacity (English) | 3.38 oz. |
Color | Amber |
Diameter (English) | 2.08 in. |
Material | Glass |
Closure Color | Amber |
Mouth | Narrow |
Height (Metric) | 115 mm |
Height (English) | 4.52 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'