Learn More
WDR93 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561180
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TQGPTISVLV ERPVKHLDKT ICAVAPVPAL PGMVLIFSKN GSVCLMDVAK REIICAFAPP GAFPLEVPWK PVFAVSPDHP CFLLRGDYSH ETASTDDAGI QY Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 56%.
WDR93 Polyclonal antibody specifically detects WDR93 in Human samples. It is validated for Immunohistochemistry.Specifications
WDR93 | |
Polyclonal | |
Unconjugated | |
WDR93 | |
WD repeat domain 93; WD repeat-containing protein 93; WDR93 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56964 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P2C0 | |
WDR93 | |
Recombinant protein corresponding to Human WDR93 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only