missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR45B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550864
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total WDR45B protein.
This gene encodes a member of the WIPI or SVP1 family of WD40 repeat-containing proteins. The protein contains seven WD40 repeats that are thought to fold into a beta-propeller structure that mediates protein-protein interactions, and a conserved motif for interaction with phospholipids. The human genome contains several pseudogenes of this gene.Specifications
WDR45B | |
Polyclonal | |
Unconjugated | |
WDR45B | |
0610008N23Rik; AA408735; D16Bwg0193e; RGD1305141; WD repeat domain 45B; WD repeat domain phosphoinositide-interacting protein 3; WD repeat protein 45-like; WD repeat-containing protein 45B; WD repeat-containing protein 45-like; Wdr45 like; WDR45B; Wdr45l; WDR45-like; WDR45-like protein; WIPI3; WIPI-3; WIPI49-like protein; zgc:56002 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56270 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q5MNZ6 | |
WDR45B | |
Synthetic peptide of human WD repeat domain 45B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |