missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548912
Additional Details : Weight : 0.01000kg
Description
CUL-2 is a member of the family of human cullin genes (CUL-1, -2, -3, -4a, -4b, and -5), homologous to the S. cerevisiae cdc53 gene. CUL-1 forms a complex with human p19skp1 and F box protein p45skp2. Together with cdc53 (Known as CulA), as three subunits, they form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL-1, CUL-2 does not associate with p19skp1 and p45skp2. Instead, CUL-2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL. The function of this association remains unknown.Specifications
WDR20 | |
Polyclonal | |
Unconjugated | |
WDR20 | |
2310040A13Rik; AI549910; DMR; Protein DMR; WD repeat domain 20; WD repeat domain 20A; WD repeat-containing protein 20; WDR20; Wdr20a | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
91833 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide | |
Q8TBZ3 | |
WDR20 | |
KLH conjugated synthetic peptide between 5-32 amino acids from the N-terminal region of human WDR20. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |