Learn More
WBSCR27 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554587
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LTTRTNSSNL QYKEALEATL DRLEQAGMWE GLVAWPVDRL WTAGSWLPPS WRWYPASLPR MASSPALSTC TESG Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 41%.
This gene encodes a protein belonging to ubiE/COQ5 methyltransferase family. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.22-q11.23. [provided by RefSeq, Jul 2008]Specifications
WBSCR27 | |
Polyclonal | |
Unconjugated | |
METTL27 | |
AW492986; BC002286; Methyltransferase-like protein 27; METTL27; WBSCR27; Williams Beuren syndrome chromosome region 27; Williams Beuren syndrome chromosome region 27 (human); Williams-Beuren syndrome chromosomal region 27 protein; Williams-Beuren syndrome chromosome region 27 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
155368 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6F8 | |
METTL27 | |
Recombinant protein corresponding to Human WBSCR27 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only