Learn More
WBP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564235
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VNNTESILMS YDHVELTFND MKNVPEAFKG TKKGTVYLTP YRVIFLSKGK DAMQSFMMPF YLMKDCEIKQ PVFG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 97%.
The globular WW domain is composed of 38 to 40 semiconserved amino acids shared by proteins of diverse functions including structural, regulatory, and signaling proteins. The domain is involved in mediating protein-protein interactions through the binding of polyproline ligands. WBP2 is a WW domain binding protein, which binds to the WW domain of Yes kinase-associated protein by its PY motifs. The function of this protein has not been determined.Specifications
WBP2 | |
Polyclonal | |
Unconjugated | |
WBP2 | |
GRAMD6; Wbp2; WBP-2; WW domain binding protein 2; WW domain-binding protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969T9 | |
WBP2 | |
Recombinant protein corresponding to Human WBP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only