missing translation for 'onlineSavingsMsg'
Learn More

Water, TraceSELECT™, For Trace Analysis, Honeywell Riedel-de Haën™

Product Code. 15617550
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
10 L
4 x 2.5 L
Packaging:
Cubitainer
Glass Bottle
Poly Bottle

TraceSELECT™, for trace analysis

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 6 to 8 (at 20°C)
Color Colorless
Melting Point 0°C
Boiling Point 100°C
UN Number NONH for all modes of transport
Beilstein 2050024
Formula Weight 18.02g/mol
Grade TraceSELECT™
Odor Odorless
Packaging Poly Bottle
Density 1g/mL at 20°C
Quantity 2.5 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Identifier
  • Water
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'