Learn More
WAPL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558149
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TTQHDKSGEW QETSGEIQWV STEKTDGTEE KHKKEEEDEE LDLNKALQHA GKHMEDCIVA SYTALLLGCL CQESPINVTT VREYLPEGDF SIM Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
Transcriptional repressor which may play a role in development of the central nervous system (CNS).Specifications
WAPL | |
Polyclonal | |
Unconjugated | |
WAPL | |
A530089A20Rik; BC037674; DIF-2; Dioxin-inducible factor 2; FOE; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); friend of EBNA2 protein; KIAA0261; RGD1308960; WAPAL; WAPL; WAPL cohesin release factor; wings apart-like homolog (Drosophila); wings apart-like protein homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23063 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5K2 | |
WAPL | |
Recombinant protein corresponding to Human WAPL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only