missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Reagent Reservoir Cooler
Custom-fit companion for VistaLab 100mL reagent reservoirs
Brand: VistaLab™ Technologies 3054-5000
Includes: Three 100mL reservoirs and lids.
Description
- Reusable cooler maintains fluid temperatures at less than 5°C for up to two hours
- Constructed of heavy-duty PVC that encases a quick-freezing, nontoxic bright blue gel
- Provides consistent cooling without contamination risk or inconvenience of ice baths
Specifications
Blue | |
Non-sterile | |
6 in. | |
3/Pk. | |
8 in. | |
Reusable |
PVC | |
Reservoir Cooler | |
10 in. | |
100 mL | |
Not Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction