Learn More
Camlab™ Vinyl Labels
Brand:  Camlab™ RTP095
Code : Z1
Additional Details : Weight : 0.12780kg
Product Code. 11712108
  • POA / Pack of 1000
In Stock
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Self adhesive, smear resistant with a high degree of chemical resistance
  • Available as Hi-Low strips and Hi-Low labels
  • Both types suitable for use between -85°C to 100°C
  • Designed for use with thermal transfer and laser printers
  • Hand feeding recommended