Learn More
Ansell™ VersaTouch™ 92-210 Series Single-Use Blue Nitrile Gloves
Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions. A thinner glove with enhanced fingertip flexibility and improved overall comfort.
£4.35 - £17.40
Specifications
Exterior Finish | Fully Textured |
---|---|
Material | Nitrile |
Thickness (English) | 2.9 mil |
Thickness (Metric) | 0.07 mm |
Series | 92-210 |
Description
- Single use gloves
- Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions
- Thinner glove with enhanced fingertip flexibility and improved overall comfort
- Textured fingertips provide good comfort and tactility for sensitive, precise handling
- Latex-free: no risk of Type 1 allergies
- Approved for food contact applications
- Ideal for handling all fatty food
Applications:
- Prepared meals
- Fruit and vegetable processing
- Dairy processing
- Food distribution
- Catering
Specifications
Fully Textured | |
2.9 mil | |
92-210 |
Nitrile | |
0.07 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell™ VersaTouch™ 92-210 Series Single-Use Blue Nitrile Gloves