Learn More
Veolia Water Systems Veolia Water Systems™ Micron Ultra-Microfilter
0.05 Micron Ultra-Micro Filter (UMF) ideal for passage of particles and microorganisms in purified water systems.
Brand:  Veolia Water Systems LC109
Code : A0
Additional Details : Weight : 0.80000kg
Product Code. 11954867
  • £400.92 / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
  • For passage of particles and microorganisms in purified water systems
  • Ultra-micro water filter provides a physical barrier to the passage of particles and microorganisms in purified water systems
  • 0.05 Micron Ultra-Micro Filter (UMF)
  • Part Number : LC109
  • Compatible with MAXIMA MK1/MK2, MEDICA 30/60/100 Mk2 & 3, MEDICA PRO, MEDICA-R 30/60/100, PURELAB UHQ II & UHQ PS, PURELAB Ultra Mk1, PURELAB Ultra Mk2