missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Panel Mounting
VapLock™ Solvent Waste Manifold Caps for Panel Mounting connect to waste containers for a secure closed system
Specifications
Type | Solvent Waste Manifold |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | Solvent-Compatible Containers |
Size | Panel Mount |
Description
- Fits panel aperture of 1-1/4 in and require a 1-3/4 in diameter clearance around the aperture
- Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Specifications
Solvent Waste Manifold | |
Solvent Safety | |
Solvent-Compatible Containers | |
Panel Mount |