missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Justrite™ Containers
VapLock™ Solvent Waste Kits for Justrite™ Containers collect effluent from multiple sources while minimizing harmful solvent exhaust
Specifications
Closure Size | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
---|---|
Size | Justrite Quick-Disconnect |
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | Solvent-Compatible Containers |
Description
Easily connect to Justrite safety containers using quick disconnectsWarranty
None
Specifications
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) | |
Justrite Quick-Disconnect | |
Solvent Safety | |
Solvent-Compatible Containers |