Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, Micro Ports
VapLock™ Adapters for Exhaust Filters, Micro Ports feature various adapter configurations allowing quick and easy connection to emission sources
Brand: Cole-Parmer™ 12019-85
2.60 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.11000kg
Description
- Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
- May require additional adapters to fit your container
Services
None
Specifications
Solvent Safety | |
Filter Adapter, Luer lock x 1/4 in. NPT(M), natural polypropylene; 1/ea | |
VapLock Exhaust Filters |
NPT(M) x Luer Lock, Male | |
White Polypropylene (PP) | |
1/4 in. NPT(M) |