missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Filter Assemblies for High Performance Transilluminators
£555.00 - £1240.00
Product Code | Brand | Filtration Area (English) | Filtration Area (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filtration Area (English) | Filtration Area (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
12862999
|
UVP
38-0190-11 |
9.84 x 11.81 in. | 25 x 30 cm | TFM-30, TFS-30, TFML-30 high performance transilluminator |
£1150.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
12852999
|
UVP
38-0190-09 |
8.26 x 10.23 in. | 21 x 26 cm | TFM-26, TFS-26, TFML-26 high performance transilluminator |
£1100.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
12832999
|
UVP
38-0190-10 |
7.87 x 15.74 in. | 20 x 40 cm | TFL-40 high performance transilluminator |
£560.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
12872999
|
UVP
38-0190-04 |
7.87 x 15.74 in. | 20 x 40 cm | TFM-40, TFS-40, TFML-40 high performance transilluminator |
£1240.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
12842999
|
UVP
38-0190-02 |
7.87 x 7.87 in. | 20 x 20 cm | TFM-20, TFS-20, TFML-20 high performance transilluminator |
£555.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
Specifications
Filter Assembly |