man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Gel Electrophoresis Equipment and Supplies
Gel Electrophoresis Equipment
Gel Imaging and Documentation Systems
UVP Clear UV Transmitting Covers
Print
UVP Clear UV Transmitting Covers
£107.00
See promotional offers below
Specifications
Color
Clear
Product Type
UV Transmitting Cover
View More Specs
Products
1
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
1
Product Code
Brand
For Use With (Equipment)
Price
Quantity & Availability
Product Code
Brand
For Use With (Equipment)
Price
Quantity & Availability
12843848
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
UVP
19-0140-01
For use with xx-15 series benchtop uv lamps
£107.00
Each
View alternative products
Save Up to
»
Estimated Shipment: 05-08-2024
Log in
to see stock.
Add to basket
Description
Description
Specifications
Specifications
Color
Clear
Product Type
UV Transmitting Cover
Videos
SDS
Documents
Documents
Product Certifications
Back to Top