missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP44 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£122.57 - £303.85
Specifications
Antigen | USP44 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16814734
|
Proteintech
15521-1-AP-20UL |
20 μL |
£122.57
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16804734
|
Proteintech
15521-1-AP-150UL |
150 μL |
£303.85
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP44 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.[supplied by OMIM, Mar 2008].Specifications
USP44 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q8C2S0, Q9H0E7 | |
327799, 84101 | |
USP44 Fusion Protein Ag7888 | |
Primary | |
-20°C | |
USP44 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 44, Ubiquitin thiolesterase 44, USP44 | |
Usp44 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |