missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uracil, 99+%

Catalog Number p-7022683
29.45 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
50 g
250 g
1000 g
66-22-8
C4H4N2O2
112.09
MFCD00006016
ISAKRJDGNUQOIC-UHFFFAOYSA-N
uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
1174
CHEBI:17568
1H-pyrimidine-2,4-dione
O=C1NC=CC(=O)N1
66-22-8
C4H4N2O2
112.09
MFCD00006016
ISAKRJDGNUQOIC-UHFFFAOYSA-N
uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
1174
CHEBI:17568
1H-pyrimidine-2,4-dione
O=C1NC=CC(=O)N1

Uracil is produced by hydrolysis of nucleic acid and used in biochemical research. It serves as a allosteric regulator and coenzyme for reactions in the human body and in plants. It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug. Further, it is used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uracil is produced by hydrolysis of nucleic acid and used in biochemical research. It serves as a allosteric regulator and coenzyme for reactions in the human body and in plants. It reacts with fluorine to get 5-fluorouracil, which is an anticancer drug. Further, it is used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.

Solubility
Soluble in water, aqueous ammonia, 1M sodium hydroxide solution and alkalies. Insoluble in ethanol and ether.

Notes
Incompatible with water, air and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∽330°C
Assay Percent Range ≥99%
Quantity 50 g
Beilstein 606623
Merck Index 14,9850
Solubility Information Soluble in water,aqueous ammonia,1M sodium hydroxide solution and alkalies. Insoluble in ethanol and ether.
Formula Weight 112.09
Percent Purity ≥99%
Chemical Name or Material Uracil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'