missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Touchless-90 Light Gray

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3012000133724

Additional Details : Weight : 453.40921kg

 View more versions of this product

Product Code. 15992726

  • £4584.50 / Each
Estimated Shipment: 08-11-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Space saving folding doors: less required space in front of the cabinet while doors are open
  • Effortless use: the sensor detects a person within approx. 2 seconds
  • User-friendly and safe: after detection, the folding doors open automatically within approx. 5 seconds. The sensor recognises the person while operating and hold the folding doors open
  • Always closed: if the person leaves the sensor-controlled detection area, the folding doors close automatically after approx. 30 seconds
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing  locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1742 mm
Safety storage cabinet
1050 mm
Powder-coated plain
1196 mm
504 kg
Light Gray
1968 mm
S90.196.120.FDAO
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
4 x drawer (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
524 mm
Steel
616 x 1196 x 1968 mm
524 x 1050 x 1742 mm
616 mm
Folding, Automatic opening
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers