Learn More
DuPont™ Tychem™ NT430
Product Code. 16320466 Shop All DuPont Products
Small, 7
Click to view available options
Size:
2X-Large, 11
Large, 9
Medium, 8
Small, 7
X-Large, 10
X-Small, 6
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Very comfortable hybrid nitrile. Ideal for working in damp or greasy environments and for small parts handling. Approved for food handling.
  • 0.23 mm nitrile gloves for jobs requiring chemical performance plus optimum fit, feel and comfort
  • Lightweight,'Second-Skin' feel and low-modulus formulation reduces hand fatigue
  • CE CAT III - EN ISO 374-1:2016/TYPE A, JKOPT
  • EN ISO 374-5:2016
  • Food contact compatible
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) CE CAT III
Color Blue
Material Nitrile
Size Small, 7
Thickness (English) 9 mil
Thickness (Metric) 0.23 mm
Model D15536294
  1. Store away from light and humidity
  2. Rinse gloves in running water before removing, using a neutral detergent if necessary
  3. Gloves should not be used around heat, open flame, sparks or in potentially flammable environments
  4. For Professional Use Only
Product Content Correction
Product Title
DuPont™ Tychem™ NT430 >