Learn More
DuPont™ Tychem™ NT430
Click to view available options
Size:
2X-Large, 11
Large, 9
Medium, 8
Small, 7
X-Large, 10
X-Small, 6
Description
- 0.23 mm nitrile gloves for jobs requiring chemical performance plus optimum fit, feel and comfort
- Lightweight,'Second-Skin' feel and low-modulus formulation reduces hand fatigue
- CE CAT III - EN ISO 374-1:2016/TYPE A, JKOPT
- EN ISO 374-5:2016
- Food contact compatible
Specifications
Specifications
Certifications/Compliance (Europe) | CE CAT III |
Color | Blue |
Material | Nitrile |
Size | X-Small, 6 |
Thickness (English) | 9 mil |
Thickness (Metric) | 0.23 mm |
Model | D15536293 |
- Store away from light and humidity
- Rinse gloves in running water before removing, using a neutral detergent if necessary
- Gloves should not be used around heat, open flame, sparks or in potentially flammable environments
- For Professional Use Only