missing translation for 'onlineSavingsMsg'
Learn More

Tungsten dichloride dioxide, 99%, Thermo Scientific Chemicals

Catalog Number p-4904563
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
13520-76-8
WO2Cl2
MFCD00054136
This item is not returnable. View return policy
13520-76-8
WO2Cl2
MFCD00054136

Tungsten dichloride dioxide is used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 99%
Solubility Information Soluble in cold water. Decomposes in hot water.
Formula Weight 286.75
Odor Acrid
Sensitivity Moisture sensitive
Chemical Name or Material Tungsten dichloride dioxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'