missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Tube Block
570.00 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Use promo code "21657" to get your promotional price.
Click to view available options
Holds:
Conical Tubes
Cryotubes
Eppendorf Tubes
Tubes
No. of Places:
4
8
9
24
30
Tube Size:
0.5 mL
1.5 mL
15 mL
2 mL
5 mL
5 to 7 mL
50 mL
Description
- Designed to securely hold tubes in place
- Applications include: cell cultures, bacterial and yeast cultures, immunoassays, protein studies, blotting techniques, staining and destaining gels. solubility studies, extraction procedures, DNA, RNA, hybridizations, enzyme reactions, ELISA assays
Specifications
Description | Block For 30 X 0.5 mL Tubes |
For Use With (Equipment) | Shaker |
Holds | Tubes |
Material | Valox 357 Plastic |
No. of Places | 30 |
Type | Block |
Tube Size | 0.5 mL |
Dimensions (L x W) | 10.20 cm x 14.20 cm |
Dimensions (L x W x H) | 10.2 x 14.2 x 4.6 cm |
Length (English) | 4.00 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'