Learn More
Triphenylamine, 98%, Thermo Scientific Chemicals
£16.20 - £138.00
Chemical Identifiers
CAS | 603-34-9 |
---|---|
Molecular Formula | C18H15N |
Molecular Weight (g/mol) | 245.325 |
MDL Number | MFCD00003020 |
InChI Key | ODHXBMXNKOYIBV-UHFFFAOYSA-N |
Synonym | triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2 |
PubChem CID | 11775 |
IUPAC Name | N,N-diphenylaniline |
SMILES | C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448476
|
Thermo Scientific Alfa Aesar
A15939.06 |
5 g |
£16.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458476
|
Thermo Scientific Alfa Aesar
A15939.14 |
25 g |
£43.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468476
|
Thermo Scientific Alfa Aesar
A15939.22 |
100 g |
£138.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
603-34-9 | |
245.325 | |
ODHXBMXNKOYIBV-UHFFFAOYSA-N | |
11775 | |
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3 |
C18H15N | |
MFCD00003020 | |
triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2 | |
N,N-diphenylaniline |
Specifications
603-34-9 | |
365°C | |
MFCD00003020 | |
triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2 | |
ODHXBMXNKOYIBV-UHFFFAOYSA-N | |
N,N-diphenylaniline | |
11775 | |
98% |
125°C to 129°C | |
C18H15N | |
2050487 | |
Insoluble in water. | |
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3 | |
245.325 | |
245.33 | |
Triphenylamine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-035-5
missing translation for 'rtecsNumber' : YK2680000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only