missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyltin chloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is used in the synthesis of some tin alkyls and also in the preparation of organolithium compounds by the transmetalation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.429g/mL |
Boiling Point | 206°C |
Odor | Unpleasant |
Quantity | 5 g |
UN Number | UN2788 |
Sensitivity | Moisture Sensitive |
Formula Weight | 241.33 |
Physical Form | Liquid |
Chemical Name or Material | Triethyltin chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'