missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-4509966
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
545-06-2
C2Cl3N
144.39
DRUIESSIVFYOMK-UHFFFAOYSA-N
trichloroacetonitrile, acetonitrile, trichloro, tritox, cyanotrichloromethane, trichloromethyl cyanide, trichloroethanenitrile, trichlouracetonitril, trichlor-acetonitrile, trichloromethylnitrile, nitrile trichloracetique
11011
CHEBI:82541
2,2,2-trichloroacetonitrile
C(#N)C(Cl)(Cl)Cl
This item is not returnable. View return policy
545-06-2
C2Cl3N
144.39
DRUIESSIVFYOMK-UHFFFAOYSA-N
trichloroacetonitrile, acetonitrile, trichloro, tritox, cyanotrichloromethane, trichloromethyl cyanide, trichloroethanenitrile, trichlouracetonitril, trichlor-acetonitrile, trichloromethylnitrile, nitrile trichloracetique
11011
CHEBI:82541
2,2,2-trichloroacetonitrile
C(#N)C(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -42.0°C
Color Colorless to Yellow
Density 1.4400g/mL
Boiling Point 83.0°C to 84.0°C
Flash Point 195°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CCl3CN
Refractive Index 1.4400 to 1.4420
Quantity 5 g
Beilstein 02,212
Fieser 01,1194; 05,686; 06,604; 07,381; 12,526; 13,32
Merck Index 15,9793
Specific Gravity 1.44
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Formula Weight 144.39
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trichloroacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'