missing translation for 'onlineSavingsMsg'
Learn More

tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane, Thermo Scientific™

£58.70 - £480.00

Chemical Identifiers

CAS 1007847-70-2
Molecular Formula C22H39NO2Sn
Molecular Weight (g/mol) 468.269
InChI Key TYKSDTUCXMRYNR-UHFFFAOYSA-N
Synonym 2-oxan-4-yloxy-6-tributylstannyl pyridine, tributyl 6-tetrahydropyran-4-yloxy pyrid-2-yl stannane, 2-oxan-4-yl oxy-6-tributylstannyl pyridine
PubChem CID 45594295
IUPAC Name tributyl-[6-(oxan-4-yloxy)pyridin-2-yl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)OC2CCOCC2
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10205324
View Documents Promotion Details
Thermo Scientific Maybridge
CC58241CB
250 mg Amber Glass Bottle
£58.70
250mg
Please to purchase this item. Need a web account? Register with us today!
10012164
View Documents Promotion Details
Thermo Scientific Maybridge
CC58241DA
1 g Amber Glass Bottle
£116.00
1g
Please to purchase this item. Need a web account? Register with us today!
10662865
View Documents Promotion Details
Thermo Scientific Maybridge
CC58241DE
5 g Amber Glass Bottle
£480.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1007847-70-2
468.269
2-oxan-4-yloxy-6-tributylstannyl pyridine, tributyl 6-tetrahydropyran-4-yloxy pyrid-2-yl stannane, 2-oxan-4-yl oxy-6-tributylstannyl pyridine
tributyl-[6-(oxan-4-yloxy)pyridin-2-yl]stannane
C22H39NO2Sn
TYKSDTUCXMRYNR-UHFFFAOYSA-N
45594295
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)OC2CCOCC2

Specifications

1007847-70-2
2-oxan-4-yloxy-6-tributylstannyl pyridine, tributyl 6-tetrahydropyran-4-yloxy pyrid-2-yl stannane, 2-oxan-4-yl oxy-6-tributylstannyl pyridine
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)OC2CCOCC2
468.269
468.25
tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane
C22H39NO2Sn
TYKSDTUCXMRYNR-UHFFFAOYSA-N
tributyl-[6-(oxan-4-yloxy)pyridin-2-yl]stannane
45594295
90%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications