missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Triangular Magnetic Stirring Bar

Kartell™ Triangular Magnetic Stirring Bar is ideal for dissolving solids and mixing sediments thanks to its scraping action.

Brand:  Kartell™ 0078000

Code : A0

Additional Details : Weight : 0.01000kg

Product Code. 12637795

  • £30.15 / Pack of 10
Estimated Shipment: 12-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Cuneiform
  • The angled surfaces create high turbulence even at low speeds
  • Suitable for foodstuff
Specifications

Specifications

5.61mm
Magnet PTFE Coated
White
Magnet
Triangular
PTFE (Polytetrafluoroethylene)
Stirring bar
10/Pk.
12.21mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers