missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylamine, 95%, Thermo Scientific Chemicals

Product Code. 11407357
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.

Solubility
Miscible with chloroform. Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, carbon dioxide, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1116-76-3
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.679
MDL Number MFCD00009560
InChI Key XTAZYLNFDRKIHJ-UHFFFAOYSA-N
Synonym trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
PubChem CID 14227
IUPAC Name N,N-dioctyloctan-1-amine
SMILES CCCCCCCCN(CCCCCCCC)CCCCCCCC

Specifications

Melting Point -39°C
Density 0.81
Boiling Point 164°C to 168°C (0.7 mmHg)
Flash Point 158°C (316°F)
Odor Amine-like
Linear Formula [CH3(CH2)7]3N
Refractive Index 1.4485
Quantity 500 mL
UN Number UN3082
Beilstein 1210618
Sensitivity Air Sensitive
Solubility Information Miscible with chloroform. Immiscible with water.
Formula Weight 353.68
Percent Purity 95%
Chemical Name or Material Tri-n-octylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'