Learn More
Rogo Sampaic™ Transparent Glass Coverslips
Glass coverslips are transparent and don't leave fingerprints.
POA
Specifications
Shape | Square |
---|---|
Material | Soda Lime Glass |
Thickness (Metric) | #1 (0.13 to 0.16 mm) |
Description | Glass Coverslip |
Quantity | 100 |
Description
Glass coverslips are transparent and don't leave fingerprints.
- Manufactured in soda glass
- Uniform thickness of 0.13 to 0.16 mm
Specifications
Square | |
#1 (0.13 to 0.16 mm) | |
100 |
Soda Lime Glass | |
Glass Coverslip |