Learn More
Saint Gobain Life Sciences Chemware™ PFA Graduated Erlenmeyer Flask
Chemware™ PFA Graduated Erlenmeyer Flask is chemically inert for ultra-pure applications.
Brand:  Saint Gobain Life Sciences D1069076
 View more versions of this product
Product Code. 11724738
  • £183.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Graduated
  • Transparent for easy viewing
  • Non-stick PFA surface-easy to clean
  • Can be used in a microwave
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Translucent
PFA
1 Ea.
200 mL
123 mm