missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human and Mouse Apoptosis regulator Bcl-2 siRNA
Blocks transcription and translation of Apoptosis regulator Bcl-2 from Human and Mouse.
£227.54 - £336.64
Specifications
Accession Number | NP_000648 |
---|---|
Cell Lines Tested | Hct116 Colorectal Carcinoma Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | BCL2 |
Gene ID (Entrez) | 596 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079161
|
enQuireBio™
QX10-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069161
|
enQuireBio™
QX10-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000648 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
596 | |
Bcl-2, PPP1R50 | |
Lyophilized |
Hct116 Colorectal Carcinoma Cells. | |
BCL2 | |
siRNA | |
Human, Mouse |