missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-beta-Nitrostyrene, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
trans-beta-Nitrostyrene is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
Specifications
Melting Point | 55°C to 58°C |
Boiling Point | 250°C to 260°C |
Quantity | 5 g |
Beilstein | 1210066 |
Solubility Information | Insoluble in water. |
Formula Weight | 149.15 |
Percent Purity | 98% |
Chemical Name or Material | trans-beta-Nitrostyrene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'