missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hydroxycinnamic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081669
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
614-60-8
C9H8O3
164.16
MFCD00004379
PMOWTIHVNWZYFI-AATRIKPKSA-N
2-hydroxycinnamic acid, o-coumaric acid, trans-2-hydroxycinnamic acid, 2-coumaric acid, trans-o-hydroxycinnamic acid, trans-o-coumaric acid, 2-coumarate, e-o-hydroxycinnamic acid, 2-hydroxycinnamate, o-hydroxy-trans-cinnamic acid
637540
CHEBI:18125
(E)-3-(2-hydroxyphenyl)prop-2-enoic acid
OC(=O)\C=C\C1=CC=CC=C1O
This item is not returnable. View return policy
614-60-8
C9H8O3
164.16
MFCD00004379
PMOWTIHVNWZYFI-AATRIKPKSA-N
2-hydroxycinnamic acid, o-coumaric acid, trans-2-hydroxycinnamic acid, 2-coumaric acid, trans-o-hydroxycinnamic acid, trans-o-coumaric acid, 2-coumarate, e-o-hydroxycinnamic acid, 2-hydroxycinnamate, o-hydroxy-trans-cinnamic acid
637540
CHEBI:18125
(E)-3-(2-hydroxyphenyl)prop-2-enoic acid
OC(=O)\C=C\C1=CC=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼210°C (decomposition)
Quantity 25 g
Beilstein 1100900
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material trans-2-Hydroxycinnamic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'