missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53INP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17872-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
In response to double-strand DNA breaks, promotes p53/TP53 phosphorylation on ′Ser-46′ and subsequent apoptosis.Specifications
TP53INP1 | |
Polyclonal | |
Unconjugated | |
TP53INP1 | |
p53DINP1, SIP, Stress induced protein, Teap, TP53DINP1, TP53INP1, TP53INP1A, TP53INP1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297822, 94241 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80YE2, Q96A56 | |
TP53INP1 | |
TP53INP1 Fusion Protein Ag12358 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |