missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNFR1 (Soluble) Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EMTNFRSF1A

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15435343

  • £504.00 / 1 set
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.

Description

Description

The Mouse soluble TNFRI (TNFRSF1A) ELISA quantitates Ms TNFRI in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms sTNFRI. Principle of the method The Mouse TNF-RI solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tumor necrosis factor receptor 1 (TNFR1) belongs to the tumor necrosis factor superfamily, and is one of the major TNF-alpha receptors. This receptor plays an important role in mediating apoptosis. TNFR1 also plays an important role in cytokine mediated signaling, positive regulation of the NF-Kb pathway, positive regulation of angiogenesis, and negative regulation of gene expression. The extracellular domain of the receptor is also released into the circulatory system as soluble TNFR1 (sTNFR1). In humans, the TNFR1 gene is located on chromosome 12.
Specifications

Specifications

P25118
1 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
21937
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Inflammation
2°C to 8°C
Mouse
4 hr. 45 min.
1.23-300 pg/mL
1pg/mL to 300pg/mL
Biotin
Cytokines & Receptors
This ELISA pair antibody detects mouse sTNFRI. Other species not determined.
Mouse
Colorimetric Microplate Reader
Tnfrsf1a
<10%
HRP
RUO
Plasma, 20 μL; Serum, 20 μL; Supernatant, 50 μL
CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI, TNFRp55, TNFalpha-R1, Tnfr-2, Tnfr1, p55, p55-R
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers