missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Mouse anti-Human, Mouse, Clone: 3G9A4, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | TLR4 |
---|---|
Clone | 3G9A4 |
Concentration | 0.77 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16848145
|
Proteintech
66350-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16838145
|
Proteintech
66350-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TLR4 is a member of the Toll like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. It has been implicated in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria.Specifications
TLR4 | |
0.77 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARMD10, CD284, hToll, TLR4, TOLL, toll like receptor 4 | |
TLR4 | |
IgG2b | |
Protein A | |
Antibody |
3G9A4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human | |
O00206, Q9QUK6 | |
21898, 7099 | |
TLR4 Fusion Protein Ag13857 | |
Primary | |
-20°C | |
TLR4 |