missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE3 Mouse anti-Human, Clone: 1D11A8, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | TLE3 |
---|---|
Clone | 1D11A8 |
Concentration | 0.61 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877635
|
Proteintech
66083-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867635
|
Proteintech
66083-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.Specifications
TLE3 | |
0.61 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ESG, ESG3, GRG3, HsT18976, KIAA1547, TLE3 | |
TLE3 | |
IgG1 | |
Protein G | |
Antibody |
1D11A8 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q04726 | |
7090 | |
TLE3 Fusion Protein Ag17906 | |
Primary | |
-20°C | |
TLE3 |